Tyrphostin AG 1478 ≥98% (by HPLC)
Dodavatel: ENZO LIFE SCIENCES
Synonyms:
N-(3-Chlorophenyl)-6,7-dimethoxy-4-quinazolinamine, N-(3-Chlorophenyl)-6,7-dimethoxyquinazolin-4-amine
ALX270036M005
ENZOALX270036M005EA
4540
CZK
ENZOALX270036M005
Tyrphostin AG 1478 ≥98% (by HPLC)
Tyrphostin AG 1478
EGFR inhibitor
Potent and selective inhibitor of EGFR (IC50 = 3nM). Reduces EGF-stimulated DNA synthesis in Rat-1 fibroblasts by ~75% at 0.25μM. Blocks EGF-dependent src-family kinase activation and p21/Cip 1/WAF1 induction in A431 cells. Also inhibits H2O2-induced stimulation of amphiregulin expression in rat gastric epithelium (RGM 1) cells.
Vzorec:
C₁₆H₁₄ClN₃O₂ MW: 315,76 g/mol |
MDL Number:
MFCD00270914 CAS číslo: 153436-53-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...