Tisk…

DL-threo-1-Phenyl-2-palmitoylamino-3-morpholino-1-propanol ≥98% (by TLC)

Dodavatel: ENZO LIFE SCIENCES

Synonyms: DL-PPMP

BMLSL2150005 BMLSL2150025
ENZOBMLSL2150005EA 1950 CZK
ENZOBMLSL2150005 ENZOBMLSL2150025
DL-threo-1-Phenyl-2-palmitoylamino-3-morpholino-1-propanol ≥98% (by TLC)
DL-threo-1-Phenyl-2-palmitoylamino-3-morpholino-1-propanol

Inhibits glucosylceramide synthetase. More effective than PDMP in intact cells.


  • Glucosylceramide synthetase inhibitor


White-pink solid.


Soluble in DMSO (10mg/ml).

Vzorec: C₂₉H₅₀N₂O₃·HCl
Storage Temperature: Freezer
CAS číslo: 149022-18-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43481e91a70316b05ef52a0bbd8f0e9f90a675cb&vid=b10255ec179227f27bfc42213b326e6122fd80e7&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR