Tisk…

MBCQ ≥98%

Dodavatel: ENZO LIFE SCIENCES

BMLPD1490005 BMLPD1490025
ENZOBMLPD1490005EA 2540 CZK
ENZOBMLPD1490005 ENZOBMLPD1490025
MBCQ ≥98%
MBCQ

Selective inhibitor of cGMP-specific phosphodiesterase (PDE V, IC50=19 nM).


  • PDE inhibitor


White solid.


Soluble in DMSO.

Vzorec: C₁₆H₁₂CIN₃O₂
Storage Temperature: Ambient
CAS číslo: 150450-53-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7de405c2bfc185f39e34bf80cbbddca75109c34f&vid=5066401ea833081f3cd7d485f02851ecd63b4550&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR