Tisk…
(±)-10-Hydroxycamptothecin ≥97%
Dodavatel: ENZO LIFE SCIENCES
Synonyms: 1-HCPT, 10-Hydroxycamptothecin, 4-Ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, NSC107124
BMLGR3160020 BMLGR3160100
ENZOBMLGR3160020EA 3250 CZK
ENZOBMLGR3160020 ENZOBMLGR3160100
(±)-10-Hydroxycamptothecin ≥97%
(±)-10-Hydroxycamptothecin
A DNA topoisomerase I inhibitor that induces differentiation in HL-60 cells. It induces apoptosis in human breast cancer cell lines MCF-7 and MDA-MB-468.
  • Topoisomerase inhibitor
Pale yellow solid.
Soluble in DMSO.
Vzorec: C₂₀H₁₆N₂O₅
MW: 364.35 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09027971
CAS číslo: 64439-81-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80a3bacbeb95cd3f8e6cbaae980b109b3f52d716&vid=170ff37013d543efda901cc7a36fcc7f77503c63&prid=web&clid=SAPDAM