Tisk…

Lycorine HCl ≥97%

Dodavatel: ENZO LIFE SCIENCES

Synonyms: (1S,2S,3a1S,12bS)-2,3a1,4,5,7,12b-Hexahydro-1H-[1,3]dioxolo[4,5-j]pyrrolo[3,2,1-de]phenanthridine-1,2-diol hydrochloride, Lycorine hydrochloride

BMLGR3130010 BMLGR3130050
ENZOBMLGR3130010EA 3140 CZK
ENZOBMLGR3130010 ENZOBMLGR3130050
Lycorine HCl ≥97%
Lycorine HCl

Lycorine is a eukaryotic termination inhibitor.


  • Protein synthesis inhibitor


White to off-white powder.


Soluble in water (>15mg/ml).

Vzorec: C₁₆H₁₇NO₄·HCl
MW: 323,8 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00243111
CAS číslo: 2188-68-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=112fe242c9c173f1299f33acf3ddffd3994febb4&vid=a5c4d1238478138c3c7877549f1cb46954c16f3a&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR