Tisk…

5-Iodotubercidin ≥98% (by TLC)

Dodavatel: ENZO LIFE SCIENCES

Synonyms: 4-Amino-5-iodo-7-(β-D-ribofuranosyl)-pyrrolo[2,3-d]pyrimidine, (2R,3R,4S,5R)-2-(4-Amino-5-iodo-7H-pyrrolo[2,3-d]pyrimidin-7-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol

BMLEI2930001 BMLEI2930005
ENZOBMLEI2930001EA 3740 CZK
ENZOBMLEI2930001 ENZOBMLEI2930005
5-Iodotubercidin ≥98% (by TLC)
5-Iodotubercidin

Inhibits ERK2 (Ki = 525nM). Also inhibits adenosine kinase (Ki = 30nM), CK1 and CK2 and insulin receptor kinase (IC50 ranging from 0.4-28µM).


  • Protein kinase inhibitor


Off-white solid


Soluble in DMSO (25mg/ml) or 100% ethanol (8mg/ml warm)

Vzorec: C₁₁H₁₃IN₀₄
Storage Temperature: Freezer
MDL Number: MFCD00055131
CAS číslo: 24386-93-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb27b21f0866c77e16549284b1b8ddecd8a38e6c&vid=f4272885a9418abbfee0a4e55cfeb0e2674011d7&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR