Tisk…

Shikonin ≥98% (by HPLC)

Dodavatel: ENZO LIFE SCIENCES

BMLCT1150010 BMLCT1150050
ENZOBMLCT1150010EA 2480 CZK
ENZOBMLCT1150010 ENZOBMLCT1150050
Shikonin ≥98% (by HPLC)
Shikonin

Shikonin inhibits TNF-α- and B6 melanoma-induced angiogenesis in mice. It blocks expression of integrin αvβ3 and inhibits endothelial cell proliferation and migration in vitro.


  • Inhibits angiogenesis


Red crystalline solid.


Soluble in DMSO.

Vzorec: C₁₆H₁₆O₅
Storage Temperature: Refrigerator
MDL Number: MFCD00075680
CAS číslo: 517-89-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77792fe370b238205c892e29eddeb846edadc609&vid=7934dcc7c63d19596c3f5ff6596142cbb0d516a6&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR