Tisk…
QS11 ≥98% (by HPLC)
Dodavatel: ENZO LIFE SCIENCES
BMLWN1170005 BMLWN1170025
ENZOBMLWN1170005EA 4760 CZK
ENZOBMLWN1170005 ENZOBMLWN1170025
QS11 ≥98% (by HPLC)
QS11
Wnt pathway activator
Synergizers with Wnt3a to activate the Wnt/β-catenin pathway. Binds and inhibits GTPase activating protein of ADP-ribosylation factor 1 (ARFGAP1), thus activating ARF. It has also been suggested that a possible mechanism by which QS11 synergizes Wnt is through ARFGAP1 inhibition leading to increased β-catenin translocation via ARF activation.
Vzorec: C₃₅H₃₃N₅O₂
MW: 567.7 g/mol
MDL Number: MFCD18086864
CAS číslo: 944328-88-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00ffea57a0daea5f534783567c4c0463cd275cdc&vid=4f493aad711b8b8b7ca357a5e8fa8014217871ef&prid=web&clid=SAPDAM