Tisk…

(±)-Blebbistatin ≥98%

Dodavatel: ENZO LIFE SCIENCES

BMLEI3150005 BMLEI3150025
ENZOBMLEI3150005EA 3740 CZK
ENZOBMLEI3150005 ENZOBMLEI3150025
(±)-Blebbistatin ≥98%
(±)-Blebbistatin

Inhibitor of non-muscle myosin II


Blebbistatin (so named because of its ability to block cell blebbing) is a selective and high-affinity (IC50 approx. 4 µM) inhibitor of non-muscle myosin II. During cell division blebbistatin inhibits contraction of the cleavage furrow without disrupting mitosis. It inhibits pancreatic adenocarcinoma cellular invasiveness and PDGF-stimulated fibroblast-collagen matrix contraction.

Vzorec: C₁₈H₁₆N₂O₂
MDL Number: MFCD04113775
CAS číslo: 674289-55-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31ca389dbe4898ab893222576a6838f8ad8435d4&vid=d5829c16d9bbb59770a74b85086a93187cc5ae09&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR