Tisk…
IQ-1 ≥98% (by TLC)
Dodavatel: ENZO LIFE SCIENCES
Synonyms: 2-((4-Acetylphenyl)diazenyl)-2-(3,3-dimethyl-3,4-dihydroisoquinolin-1(2H)-ylidene)acetamide, Wnt Pathway Activator III, IQ 1, 2-(4-Acetylphenylazo)-2-(3,3-dimethyl-3,4-dihydro-2H-isoquinolin-1-ylidene)-acetamide
BMLWN1150005 BMLWN1150025
ENZOBMLWN1150005EA 3600 CZK
ENZOBMLWN1150005 ENZOBMLWN1150025
IQ-1 ≥98% (by TLC)
IQ-1
Disrupts Wnt signaling
IQ-1 in conjunction with Wnt3a has been shown to be sufficient to maintain murine embryonic stem cell proliferation in the absence of serum. Binds to the PR72/PR130 subunit of protein phosphatase PP2A preventing β-catenin from switching co-activator usage from CBP to p300 leading to an increase in β-catenin/CBP-driven transcription at the expense of β-catenin/p300-driven transcription. Upregulates expression of Oct4 and Sox2 transcription factors.
Vzorec: C₂₁H₂₂N₄O₂
MW: 362,43 g/mol
MDL Number: MFCD00354369
CAS číslo: 331001-62-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=943705bcc8344714bca0b0712d2ddb17124def6d&vid=3bec1fe4046262f4c8d4a7a457bad7b7e7e504d0&prid=web&clid=SAPDAM