Tisk…
AGK7 ≥99% (by HPLC)
Dodavatel: ENZO LIFE SCIENCES
ENZCHM1460005 ENZCHM1460010
ENZOENZCHM1460005EA 4080 CZK
ENZOENZCHM1460005 ENZOENZCHM1460010
AGK7 ≥99% (by HPLC)
AGK7
Selectively inhibits sirtuin 3 over sirtuins 1 and 2.
Can be used as an inactive control in experiments with AGK2.
MW: 434.3 g/mol
CAS číslo: 304896-21-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=953e981d46ba7ada3ae4cafc9a07e153f3bc2554&vid=3515e5f59ca9a8bec212f7035b2959db486f4529&prid=web&clid=SAPDAM