Tisk…
Cantharidic acid ≥98% (by TLC)
Dodavatel: ENZO LIFE SCIENCES
BMLPR1060010 BMLPR1060050
ENZOBMLPR1060010EA 2480 CZK
ENZOBMLPR1060010 ENZOBMLPR1060050
Cantharidic acid ≥98% (by TLC)
Cantharidic acid
PP2A inhibitor
Potent and selective inhibitor of PP2A, IC50=53nM (IC50 for PP1=562nM). Useful for inhibiting PP1 and PP2A during purification of phosphorylated proteins.
Vzorec: C₁₀H₁₄O₅
MW: 214.22 g/mol
MDL Number: MFCD00047137
CAS číslo: 28874-45-5