Tisk…
Ro 41-5253 ≥98% (by TLC)
Dodavatel: ENZO LIFE SCIENCES
BMLGR1100005 BMLGR1100025
ENZOBMLGR1100005EA 3450 CZK
ENZOBMLGR1100005 ENZOBMLGR1100025
Ro 41-5253 ≥98% (by TLC)
Ro 41-5253
Retinoic acid receptor ligand
A selective RARα antagonist (concentration required to inhibit 50% of specific retinoic acid binding: RARα, 60 nM; RARß, 2400 nM; RARτ, 3300 nM). Counteracts the effects of retinoic acid on HL-60 cell differentiation, B-lymphocyte polyclonal activation and in a RARα transactivation assay. Ro 41-5253 suppresses retinoic acid-induced connexin 43 protein expression in 10T1/2 cells.
MW: 484.7 g/mol
CAS číslo: 144092-31-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bad28e3fe5e30c98a1ff3bdd357f563c724816df&vid=6713307e4ce96a7bced340a092d48fea4a3f85a1&prid=web&clid=SAPDAM