Tisk…
WYE-23 ≥99% (by HPLC)
Dodavatel: ENZO LIFE SCIENCES
ENZCHM1360001 ENZCHM1360005
ENZOENZCHM1360001EA 3850 CZK
ENZOENZCHM1360001 ENZOENZCHM1360005
WYE-23 ≥99% (by HPLC)
WYE-23
Cell-permeable, potent, selective, and ATP-competitive mTOR inhibitor.
MW: 520,6 g/mol
CAS číslo: 1062169-46-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b5bc731d9af6a9f758292256e36ea3d4ffefe58&vid=0856d0d8c3f0066ad37e8d3ddbac0f34a171f94b&prid=web&clid=SAPDAM