Histone Deacetylase Inhibitor VIII ≥98% (by TLC)
Dodavatel: ENZO LIFE SCIENCES
Synonyms:
tert-Butyl (4-(3-((7-(hydroxyamino)-7-oxoheptyl)carbamoyl)isoxazol-5-yl)phenyl)carbamate, BML-281, CAY10603, HDAC6 Inhibitor
HDAC6 inhibitor
Selective HDAC6 inhibitor (IC50=0.002nM). IC50 values for other HDACs are 271, 252, 0.42, 6851 and 90.7 nM for HDACs 1,2,3,8 and 10 respectively. Potent inhibitor of the proliferation of a variety of pancreatic cancer cell lines (IC50s in the 0.1-0.6µM range).
Vzorec:
C₂₂H₃₀N₄O₆ MW: 446.5 g/mol |
MDL Number:
MFCD17010286 CAS číslo: 1045792-66-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...