Tisk…

AM 580 ≥98% (by TLC)

Dodavatel: Enzo Life Sciences

BMLGR1040005 BMLGR1040025
ENZOBMLGR1040005EA 4710 CZK
ENZOBMLGR1040005 ENZOBMLGR1040025
AM 580 ≥98% (by TLC)
AM 580

Retinoic acid analog which acts as a selective RARα agonist with no affinity for RXRs. Induces differentiation in leukemia cell lines and inhibits angiogenesis in chick embryos.


  • Retinoic acid receptor ligand


White to off-white solid.


Soluble in DMSO (25mg/ml) and ethanol (10mg/ml).

Vzorec: C₂₂H₂₅NO₃
Storage Temperature: Freezer
MDL Number: MFCD00673916
CAS číslo: 102121-60-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbca291264cfaa35adf2204220afbb5b6c286be4&vid=c6d9ccedeac2847b0a7ee3738d0bcc622bd166b9&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR