Triacsin C 1mg/ml ≥90% (by TLC)
Dodavatel: ENZO LIFE SCIENCES
Acyl-CoA synthetase inhibitor
Analog of polyunsaturated fatty acid. Potent inhibitor of long-chain fatty acyl CoA synthetase. Selectively inhibits arachidonoyl-CoA synthetase in intact cells and the nonspecific acyl-CoA synthetase in cell sonicates IC50=3.6-8.7µM. Inhibits neutrophil functions. Blocks β-cell apoptosis induced by fatty acids (lipoapoptosis) and synthesis of triglycerides, diglycerides and cholesterol esters. Potent vasodilator.
Vzorec:
C₁₁H₁₇N₃O |
CAS číslo:
76896-80-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...