Tisk…

Herbimycin A ≥98% (by HPLC)

Dodavatel: ENZO LIFE SCIENCES

BMLEI2270100 BMLEI2271000
ENZOBMLEI2270100EA 6020 CZK
ENZOBMLEI2270100 ENZOBMLEI2271000
Herbimycin A ≥98% (by HPLC)
Herbimycin A

Antibiotic. Cell permeable tyrosine kinase inhibitor. It is effective on Src, Yes, Fps, Ros, Abl and ErbB oncogene products and inhibits thrombin-induced tyrosine phosphorylation of phospholipase C. It also inhibits HSP90, NF-κB activation and angiogenesis. Increases the sensitivity of certain cancer cells to chemotherapeutic agents. Blocks mitotic activity. Neurotoxic.


  • White to off-white lyophilized solid


Soluble in ethanol, methanol, DMF or DMSO

Vzorec: C₃₀H₄₂N₂O₉
Storage Temperature: Freezer
MDL Number: MFCD00151702
CAS číslo: 70563-58-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=000bfe9323056f9341af98ef553990c271c3fe00&vid=fe880f4ba7a546481572d59883b0699048996d75&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR