Tisk…

4-Hydroxynonenal ≥97%

Dodavatel: ENZO LIFE SCIENCES
Varování

Synonyms: 4-Hydroxy-2-nonenal, (±)4-Hydroxynon-2-enal, HNE, 4-HNE, 4-hydroxy-2(E)-nonenal, 4-Hydroxynon-2-enal

BMLHA0040001 BMLHA0040010
ENZOBMLHA0040001EA 2540 CZK
ENZOBMLHA0040001 ENZOBMLHA0040010
4-Hydroxynonenal ≥97%
4-Hydroxynonenal

Lipid peroxidation.


Cytotoxic product of microsomal lipid peroxidation induced by xenobiotics. Reported to be a specific chemical indicator of lipid peroxidation in rat liver microsomes and in myocardial reperfusion injury. Covalently reacts with LDL and HDL during oxidative modification. Modifies proteins by covalent reaction with histidine and lysine residues. Activates TRPA1.


Soluble in 100% ethanol (25 mg/ml).

Vzorec: C₉H₁₆O₂
MW: 156,22 g/mol
Storage Temperature: Ultra-Low Temperature Freezer (–80°C)
MDL Number: MFCD00211020
CAS číslo: 75899-68-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=434d0a7d468f2e26be92fe3d487b1c58abea0462&vid=ac60a2a8529392c35bfcb5de9d76f5db6f57e088&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Purity ≥97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR