Tisk…

1-Oleoyl-2-acetyl-sn-glycerol ≥98% (by TLC)

Dodavatel: ENZO LIFE SCIENCES

BMLDG1250010 BMLDG1250050
ENZOBMLDG1250010EA 3850 CZK
ENZOBMLDG1250010 ENZOBMLDG1250050
1-Oleoyl-2-acetyl-sn-glycerol ≥98% (by TLC)
1-Oleoyl-2-acetyl-sn-glycerol

PKC activator.


In vitro potent activator of protein kinase C (PKC). Stimulator of secretion and aggregation in blood platelets. Stimulates formation of actin nucleation sites. Stimulation of pp60c-src phosphorylation in EBV transformed B-cells, 470 μM.


Soluble in DMSO or ethanol.

Vzorec: C₂₃H₄₂O₅
Storage Temperature: Freezer
CAS číslo: 86390-77-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27cef47d2ba3487cf240bfb7cf937362b0af86b7&vid=401a937f48e726b3e79b4029bd49e320543c97f4&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Source Synthetic
Purity ≥98% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR