Tisk…
Mono-n-dodecyl phosphate ≥98% (by TLC), white to off-white solid
Dodavatel: ENZO LIFE SCIENCES
Synonyms: Dihydrogen monolauryl phosphate
BMLLP1050005 BMLLP1050025
ENZOBMLLP1050005EA 3310 CZK
ENZOBMLLP1050005 ENZOBMLLP1050025
Mono-n-dodecyl phosphate ≥98% (by TLC), white to off-white solid
Mono-n-dodecyl phosphate
Lysophosphatidic acid (LPA) receptor ligand. A specific agonist at LPA2 (EC₅₀=700 nM) and antagonist at LPA.
Soluble in DMSO (15 mg/ml warm) or ethanol (2 mg/ml warm).
Vzorec: C₁₂H₂₇O₄P
MW: 266.318 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00013946
CAS číslo: 2627-35-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0670b9c2b4a74e619727e9e51c1594c979925243&vid=9347774b82b84b6e7e21debcae43023802a6419a&prid=web&clid=SAPDAM
Objednat
Výsledky specifikačního testu
Purity ≥98% (TLC)