Tisk…

4-Hydroxybenzylidenemalononitrile ≥98% (by TLC)

Dodavatel: ENZO LIFE SCIENCES

BMLPR1130010 BMLPR1130050
ENZOBMLPR1130010EA 3140 CZK
ENZOBMLPR1130010 ENZOBMLPR1130050
4-Hydroxybenzylidenemalononitrile ≥98% (by TLC)
4-Hydroxybenzylidenemalononitrile

The tyrphostins are well known tyrosine kinase inhibitors. However, tyrphostin 8 has been shown to inhibit calcineurin (IC50=21 µM) while displaying weak inhibition of EGFR tyrosine kinase (IC50=560 µM).


  • Calcineurin inhibitor


Light orange solid.


Soluble in 100% ethanol (25mg/ml) or DMSO (25mg/ml).

Vzorec: C₁₀H₆N₂O
Storage Temperature: Ambient
MDL Number: MFCD00020189
CAS číslo: 3785-90-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ab877b0a57af7aed2a0d9f7b5c2e4033d9a5b68&vid=ee8ff4ed448b8f1d93b9197b077a8896bf84cde2&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR