Tisk…

Misoprostol (free acid) ≥98% (by TLC)

Dodavatel: ENZO LIFE SCIENCES
Nebezpečí

Synonyms: (±)-15-Deoxy-(16R,S)-16-hydroxy-16-methyl-prostaglandin E1

BMLPG0520001 BMLPG0520010
ENZOBMLPG0520001EA 2420 CZK
ENZOBMLPG0520001 ENZOBMLPG0520010
Misoprostol (free acid) ≥98% (by TLC)
Misoprostol (free acid)

Prostanoid receptor ligand


Free acid form of the cytoprotective PGE₁ analog misoprostol. Misoprostol prevents NSAID-induced gastric ulceration. The free acid form is active in cell free assays and in cell culture and receptor preparations (receptor binding assays etc.). The methyl ester form (misoprostol) is a prodrug and must be hydrolyzed via intracellular or other esterases in order to display biological activity.


Soluble in 100% ethanol (>15 mg/ml) or DMSO (>20 mg/ml).

Vzorec: C₂₁H₃₆O₅
MW: 368.514 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00869984
CAS číslo: 112137-89-0
UN: 2810
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91c9c2c27e72dc5e27475092770e597b740c0760&vid=5997679357f65b89c1757a9c96717ebf5a22aa83&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Purity ≥98% (TLC)
Appearance Yellow viscous oil


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR