Tisk…

Citrinin ≥98%

Dodavatel: ENZO LIFE SCIENCES

Synonyms: Citrinin, yellow, (3S,4R)-8-hydroxy-3,4,5-trimethyl-6-oxo-4,6-dihydro-3H-isochromene-7-carboxylic acid

BMLCM1160005 BMLCM1160025
ENZOBMLCM1160005EA 3140 CZK
ENZOBMLCM1160005 ENZOBMLCM1160025
Citrinin ≥98%
Citrinin

MPTP activator


Antibiotic. Induces mitochondrial permeability pore opening and inhibits respiration by interfering with complex I of the respiratory chain. Mycotoxin. Acts as nephrotoxin in all species in which it has been tested. Has been implicated as a cause of Balkan nephropathy and yellow rice fever in humans. Induces apoptosis.


Isolated from Penicillium citrinum

Vzorec: C₁₃H₁₄O₅
MW: 250,25 g/mol
MDL Number: MFCD00006912
CAS číslo: 518-75-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0003a66f4db2b0401a2ba8ff27ff5d847cd9a44&vid=2a351bcd7ffbb539ea5f9a526d230c345a556788&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR