Fumonisin B1 ≥98%
Dodavatel: ENZO LIFE SCIENCES
Ceramide synthase inhibitor
A fungal metabolite produced by Fusarium moniliforme. Mycotoxin. Inhibits sphinganine N-acyl-transferase(ceramide synthase)(IC50=0.1µM). Inhibits de novo sphingolipid biosynthesis (IC50=0.7µM for sphingomyelin labeling by [3H]sphinganine, cultured cerebellar neurons). Inhibits growth of axons in cultured hippocampal neurons by blocking glycosphingolipid synthesis. Induces DNA damage in the liver and cancer. Induces apoptosis. Activates Akt (protein kinase B; PKB), which leads to increased survival, inhibition of GSK-3β activity and post-translational stabilization of cyclin D1.
Vzorec:
C₃₄H₅₉NO₁₅ |
MDL Number:
MFCD00133349 CAS číslo: 116355-83-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...