Tisk…

13(S)-Hydroperoxyocta-deca-9Z,11E-dienoic acid ≥98%

Dodavatel: ENZO LIFE SCIENCES

BMLHP0130050 BMLHP0131000
ENZOBMLHP0130050EA 2540 CZK
ENZOBMLHP0130050 ENZOBMLHP0131000
13(S)-Hydroperoxyocta-deca-9Z,11E-dienoic acid ≥98%
13(S)-Hydroperoxyocta-deca-9Z,11E-dienoic acid

Suppresses activation of platelets. Induces endothelial injury. Constricts isolated coronary arteries. Inhibits prostacyclin synthase.


  • Inhibits prostacyclin synthase


Suppresses activation of platelets. Induces endothelial injury. Constricts isolated coronary arteries. Inhibits prostacyclin synthase.

Vzorec: C₁₈H₃₂O₄
Storage Temperature: Ultra-Low Temperature Freezer (–80°C)
CAS číslo: 33964-75-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd01c4616b318d66e57dc79f1e780bef797cc338&vid=53dadacc07ee619452402605e6494cb67ea042a9&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR