Tisk…
N-Hexanoyl-D-erythro-sphingosine ≥98% (by TLC)
Dodavatel: ENZO LIFE SCIENCES
Synonyms: C6 ceramide, Caproyl ceramide, N-((2S,3R,E)-1,3-Dihydroxyoctadec-4-en-2-yl)hexanamide
BMLSL1100005 BMLSL1100025
ENZOBMLSL1100005EA 1890 CZK
ENZOBMLSL1100005 ENZOBMLSL1100025
N-Hexanoyl-D-erythro-sphingosine ≥98% (by TLC)
N-Hexanoyl-D-erythro-sphingosine
A biologically active, cell-permeable, but nonphysiologic ceramide analog. It activates protein phosphatase 2A and MAP kinase (MAPK/ERK), suppresses insulin-induced tyrosine phosphorylation and inhibits glycoprotein traffic by the secretory pathway. It inhibits diacylglycerol accumulation and phospholipase D activation in fibroblasts. It induces an arrest at the G0/G1 transition of the cell cycle (10μM) and causes apoptosis (15μM or greater) in Molt-4 leukemia cells.
  • Cell-permeable ceramide analog
White waxy solid.
Soluble in 100% ethanol (>25mg/ml) and DMSO (>50mg/ml).
Vzorec: C₂₄H₄₇NO₃
MW: 397.644 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00870174
CAS číslo: 124753-97-5