Tisk…

Epoprosterol sodium ≥98% (by TLC)

Dodavatel: ENZO LIFE SCIENCES

BMLPG0110001 BMLPG0110010
ENZOBMLPG0110001EA 3140 CZK
ENZOBMLPG0110001 ENZOBMLPG0110010
Epoprosterol sodium ≥98% (by TLC)
Epoprosterol sodium

Biosynthesis in endothelial cells. Induces vasodilation, inhibits platelet aggregation by receptor-mediated stimulation of adenylate cyclase. Cytoprotective. Fibrinolytic. Half-life under physiological conditions is 2 minutes.


  • Prostanoid receptor ligand


White solid.


Soluble in basic buffer (unstable at neutral or acidic pH).  The higher the pH the more stable the compound will be, but for in vitro or in vivo use  pH ~9 is recommended.  Solubility at pH 9.0 is ~10mg/ml.

Vzorec: C₂₀H₃₁NaO₅
Storage Temperature: Freezer
MDL Number: MFCD00135629
CAS číslo: 61849-14-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cb8e23dc49aef5a4dbb785f81682a8ff28118d1&vid=c9849dae1f7da90c766fe9b0f8794b076b82e847&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR