Tisk…
Clozapine-N-oxide ≥99% (by HPLC), yellow solid
Dodavatel: ENZO LIFE SCIENCES
Nebezpečí
BMLNS1050005 BMLNS1050025
ENZOBMLNS1050005EA 2970 CZK
ENZOBMLNS1050005 ENZOBMLNS1050025
Clozapine-N-oxide ≥99% (by HPLC), yellow solid
Clozapine-N-oxide
Major metabolite of clozapine. Possesses little or no activity at 5-HT receptors.
  • High Purity
  • Active in vitro and in vivo
Soluble in water (5 mg/ml).
Vzorec: C₁₈H₁₉ClN₄O
MW: 342.827 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00210190
CAS číslo: 34233-69-7
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e367c1ac802e0577184f65a9ef49fa203f5a0d22&vid=2a9a06e1acb6db4974ae2b9c14eb4451604921a0&prid=web&clid=SAPDAM
Objednat
Výsledky specifikačního testu
Purity ≥99% (HPLC)