SC-51089 ≥98%
Dodavatel: ENZO LIFE SCIENCES
BMLRA1130025
ENZOBMLRA1130025EA
12600
CZK
ENZOBMLRA1130025
SC-51089 ≥98%
SC-51089
Prostanoid receptor inhibitor
A potent and selective prostaglandin E2 (EP1) antagonist. Displays in vivo analgesic activity in the rat. Does not block PGE1-induced hyperalgesia.
Vzorec:
C₂₂H₂₀Cl₂N₄O₃ |
MDL Number:
MFCD00869867 CAS číslo: 146033-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...