Tisk…
N-(1-Adamantyl)-N'-cyclohexylmorpholine-4-carboxamidine hydrochloride ≥98%
Dodavatel: ENZO LIFE SCIENCES
Synonyms: U-37883A
BMLKC1590005 BMLKC1590025
ENZOBMLKC1590005EA 3250 CZK
ENZOBMLKC1590005 ENZOBMLKC1590025
N-(1-Adamantyl)-N'-cyclohexylmorpholine-4-carboxamidine hydrochloride ≥98%
N-(1-Adamantyl)-N'-cyclohexylmorpholine-4-carboxamidine hydrochloride
K+ channel blocker
A non-sulfonylurea inhibitor of K-ATP channels. Inhibits vascular K-ATP channels. Inhibits dopamine-modulated potassium channels.
Vzorec: C₂₁H₃₆ClN₃O
MW: 381.989 g/mol
MDL Number: MFCD03840615
CAS číslo: 57568-80-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fd2dbe52be90266bc7ac3716eaee21d1a836524&vid=28152fad05d80669fa8419561cf02abfd67bd27f&prid=web&clid=SAPDAM