Tisk…

SQ 22536 ≥99% (by TLC)

Dodavatel: ENZO LIFE SCIENCES

Synonyms: 9-(Tetrahydro-2-furyl)adenine, 9-(Tetrahydro-2-furanyl)-9H-purin-6-amine, 9-(Tetrahydrofuran-2-yl)-9H-purin-6-amine, SQ22536

BMLCN1400005 BMLCN1400025
ENZOBMLCN1400005EA 3140 CZK
ENZOBMLCN1400005 ENZOBMLCN1400025
SQ 22536 ≥99% (by TLC)
SQ 22536

Adenylate cyclase inhibitor


SQ 22536 is a cell-permeable adenylate cyclase inhibitor. It inhibits cAMP production induced by prostaglandin E1 in platelet lysates (IC50=82 µM) and in intact platelets (IC50=1 µM). It has been successfully employed as a probe for the involvement of adenylate cyclase in a variety of receptor systems including those for prostaglandins and neurotransmitters. ReducesPGE2-induced inhibition of O2- production.

Vzorec: C₉H₁₁N₅O
MDL Number: MFCD00210216
CAS číslo: 17318-31-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc0e3b331d60a0e6556154a404a18b64812cd5b8&vid=5f54399ab260b3b75fefe0dee437898ff20a17d3&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR