Tisk…

SB-366791 ≥98% (by TLC)

Dodavatel: ENZO LIFE SCIENCES
Varování

BMLVR1050005 BMLVR1050025
ENZOBMLVR1050005EA 2420 CZK
ENZOBMLVR1050005 ENZOBMLVR1050025
SB-366791 ≥98% (by TLC)
SB-366791

A potent and selective vanilloid receptor (TRPV1) antagonist. An effective antagonist of hTRPV1 when stimulated by capsaicin, acid, or noxious heat.


Soluble in DMSO (25 mg/ml) or 100% ethanol (10 mg/ml with warming).

Vzorec: C₁₆H₁₄ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD01033818
CAS číslo: 472981-92-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3bc56da941aae85eadcb33deda34da89d80e7b6c&vid=5f290e95bbd869965eb19e9a254bf60c60804dc8&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Purity ≥98% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR