Tisk…
Ar231453 ≥98% (by HPLC), yellow solid
Dodavatel: ENZO LIFE SCIENCES
Synonyms: N-(2-Fluoro-4-(methylsulphonyl)phenyl)-6-(4-(3-isopropyl-1,2,4-oxadiazol-5-yl)piperidin-1-yl)-5-nitropyrimidin-4-amine, AR 231453
BMLGP1000005 BMLGP1000025
ENZOBMLGP1000005EA 2480 CZK
ENZOBMLGP1000005 ENZOBMLGP1000025
Ar231453 ≥98% (by HPLC), yellow solid
Ar231453
GPR119 activator.
Synthetic agonist of the orphan GPR119. Although endogenous lipid amides such as oleoyl ethanolamide have been reported to be agonists at GPR119, AR231453 is the first reported high potency agonist (EC50=0.68 nM). The compound displayed in vivo activity in rodents and was active in an oral glucose tolerance test in mice following oral administration. AR231453 stimulated the release of GLP-1 via GPR119 receptors in mouse models. It was shown to be a highly selective ligand with no off-target activity across a 76 receptor and enzyme profiling panel (CEREP) and no activity against 140 known and orphan GPCRs in melanophore dispersion assays.
Soluble in DMSO (>25 mg/ml).
Vzorec: C₂₁H₂₄FN₇O₅S
MW: 505,525 g/mol
Storage Temperature: Freezer
MDL Number: MFCD14706714
CAS číslo: 733750-99-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d2db09bd18661e398f0381bc7bdc38bfc70cda3&vid=9afdcb41001a30f0fa7d109d644bbfb8267e992f&prid=web&clid=SAPDAM
Objednat
Výsledky specifikačního testu
Purity ≥98% (HPLC)
Identity Determined by NMR