Tisk…
RO 48-8071 ≥95% (by HPLC)
Dodavatel: ENZO LIFE SCIENCES
Synonyms: (4-Bromophenyl)[2-fluoro-4-[[6-(methyl-2-propen-1-ylamino)hexyl]oxy]phenyl]methanone fumarate, (4-((6-(Allyl(methyl)amino)hexyl)oxy)-2-fluorophenyl)(4-bromophenyl)methanone fumarate, RO 48-8071 fumarate
BMLEI3420010 BMLEI3420050
ENZOBMLEI3420010EA 2540 CZK
ENZOBMLEI3420010 ENZOBMLEI3420050
RO 48-8071 ≥95% (by HPLC)
RO 48-8071
Inhibits cholesterol biosynthesis at the level of 2,3-oxidosqualene cyclase (OSC). Blocks OSC and cholesterol synthesis in HepG2 cells in the nanomolar range. A novel lipid lowering agent.
  • Oxido-squalene cyclase inhibitor
Pale yellow solid.
Soluble in water (25 mg/ml)
Vzorec: C₂₃H₂₇BrFNO₂.C₄H₄O₄
MW: 564.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05865242
CAS číslo: 189197-69-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5cb77a4ad0172ef14421d20dbff445f65403837&vid=8feafbbb2a008a3b4c0c79664ccc375959114654&prid=web&clid=SAPDAM