SR-12813 ≥98%
Dodavatel: ENZO LIFE SCIENCES
BMLGR2420010
BMLGR2420050
ENZOBMLGR2420010EA
3250
CZK
ENZOBMLGR2420010
ENZOBMLGR2420050
SR-12813 ≥98%
SR-12813
PXR activator
Potent and selective PXR agonist. (EC50=200 nM for human PXR). A weak activator of rat and mouse PXR. Inhibits cholesterol biosynthesis in a number of species.
Vzorec:
C₂₄H₄₂O₇P₂ |
MDL Number:
MFCD00888473 CAS číslo: 126411-39-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...