Tisk…
Tyrphostin AG-1296 ≥98% (by TLC)
Dodavatel: ENZO LIFE SCIENCES
Synonyms: 6,7-Dimethoxy-2-phenylquinoxaline, Tyrphostin AG1296
BMLEI3030005 BMLEI3030025
ENZOBMLEI3030005EA 4590 CZK
ENZOBMLEI3030005 ENZOBMLEI3030025
Tyrphostin AG-1296 ≥98% (by TLC)
Tyrphostin AG-1296
A potent inhibitor of PDGF receptor tyrosine kinase (IC50=1 µM). Also inhibits FGF receptor tyrosine kinase and c-kit. Induces apoptosis in a small-cell lung cancer cell line (H526).
  • PDGF receptor tyrosine kinase inhibitor
Off-white to brownish solid.
Soluble in DMSO or dichloromethane.
Vzorec: C₁₆H₁₄N₂O₂
MW: 266,3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00270913
CAS číslo: 146535-11-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3351f057b02283a9b3f51c848f8893eb2920aa16&vid=df2b80a96ac69c2635452e45beee545760a55830&prid=web&clid=SAPDAM