Tisk…
Fingolimod ≥98% (by HPLC)
Dodavatel: ENZO LIFE SCIENCES
Synonyms: 2-Amino-2-(4-octylphenethyl)propane-1,3-diol hydrochloride, FTY-720 hydrochloride
BMLSL2330005 BMLSL2330025
ENZOBMLSL2330005EA 2540 CZK
ENZOBMLSL2330005 ENZOBMLSL2330025
Fingolimod ≥98% (by HPLC)
Fingolimod hydrochloride
Novel immune modulator that alters migration and homing of lymphocytes. Acts as a potent agonist of sphingosine 1-phosphate receptors (S1P) after phosphorylation by sphingosine kinase. Also inhibits cytosolic phospholipase A2 independently of sphingosine 1-phosphate receptors.
  • S1P receptor ligand
White solid.
Soluble in DMSO (10mg/ml), 100% ethanol or dimethyl formamide.
Vzorec: C₁₉H₃₄ClNO₂
MW: 343.936 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00939512
CAS číslo: 162359-56-0