Tisk…

Vinpocetine ≥98%

Dodavatel: ENZO LIFE SCIENCES

Synonyms: (41S,13aS)-Ethyl 13a-ethyl-2,3,41,5,6,13a-hexahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylate

BMLPD1850020 BMLPD1850100
ENZOBMLPD1850020EA 1890 CZK
ENZOBMLPD1850020 ENZOBMLPD1850100
Vinpocetine ≥98%
Vinpocetine

PDE inhibitor


Selective inhibitor of Ca2+/calmodulin-dependent phosphodiesterase (PDE I, IC50=20 µM).

Vzorec: C₂₂H₂₆N₂O₂
MW: 350.46 g/mol
MDL Number: MFCD00211233
CAS číslo: 42971-09-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0454b3f17ca24c6a2d61d8d4245fb4d8127e4cc&vid=1151f8daf57c76ed84137d1e30eb2ef892c7d20a&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR