Tisk…

Parthenolide ≥97% (by TLC)

Dodavatel: ENZO LIFE SCIENCES

BMLT1130050 BMLT1130250
ENZOBMLT1130050EA 2540 CZK
ENZOBMLT1130050 ENZOBMLT1130250
Parthenolide ≥97% (by TLC)
Parthenolide

HDAC inhibitor


Sesquiterpene lactone. Widely used as an herbal remedy for arthritis and migraine. Has anti-inflammatory, antisecretory and spasmolytic activity. Specifically inhibits activation of NF-kappaB by preventing the degradation of IkappaBalpha and IkappaBbeta. Inhibits activation of MAP kinase (MAPK/ERK) and generation of leukotriene B4 and thromboxane B2. Potent anticancer agent. Induces apoptosis in various cancer cell lines. Specifically inhibits histone deacetylase 1 (HDAC1) without affecting other class I/II HDACs. Inhibits tubulin carboxypeptidase (TCP) activity.


Isolated from feverfew (Chrysanthemum parthenium) leaves.

Vzorec: C₁₅H₂₀O₃
MDL Number: MFCD00134592
CAS číslo: 20554-84-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR