Tisk…

Cilostazol ≥98%

Dodavatel: ENZO LIFE SCIENCES

BMLPD1270010 BMLPD1270050
ENZOBMLPD1270010EA 2480 CZK
ENZOBMLPD1270010 ENZOBMLPD1270050
Cilostazol ≥98%
Cilostazol

PDE inhibitor


Cilostazol is a PDE3A inhibitor (0.2µM) and a unique antithrombotic agent. It inhibits cell death by activating maxi-K channels.

Vzorec: C₂₀H₂₇N₅O₂
MDL Number: MFCD00866780
CAS číslo: 73963-72-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fdce6b34ed4d9e0fda9bd2620f098b479d22b0fd&vid=4f267dd4785b1b9f89327078aa49f38fbbfa62f5&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR