Tisk…

BAY 11-7082 ≥99%

Dodavatel: ENZO LIFE SCIENCES

Synonyms: (E)-3-(p-Toluenesulphonyl)acrylonitrile, (E)-3-Tosylacrylonitrile

BMLEI2780010 BMLEI2780050
ENZOBMLEI2780010EA 3250 CZK
ENZOBMLEI2780010 ENZOBMLEI2780050
BAY 11-7082 ≥99%
BAY 11-7082

NF-kappaB inhibitor


Inhibitor of cytokine induced IκBα phosphorylation resulting thereby in a decreased expression of NF-κB and adhesion molecules. Potential anti-inflammatory agent.

Vzorec: C₁₀H₉NO₂S
MDL Number: MFCD00712162
CAS číslo: 19542-67-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d5b16b04e310dcb3f47ff5bc7c6de19183b7155&vid=51d8e1d293ad0df26da25a9b8f5b5b36a01dd8fb&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR