Tisk…
Arachidonic acid ≥99% (by TLC) (20:4, n-6)
Dodavatel: ENZO LIFE SCIENCES
Synonyms: (5Z,8Z,11Z,14Z)-Icosa-5,8,11,14-tetraenoic acid
BMLFA0030100 BMLFA0031000
ENZOBMLFA0030100EA 3250 CZK
ENZOBMLFA0030100 ENZOBMLFA0031000
Arachidonic acid ≥99% (by TLC) (20:4, n-6)
Arachidonic acid
Precursor to a large family of eicosanoids. Acts as a second messenger independent of metabolism. Retrograde messenger in long-term potentiation in the nervous system. Inhibits ras-GAP. Activator for several protein kinase C (PKC) isotypes.
  • Eicosanoid precursor and second messenger
Colorless oil.
100mg/ml soluble in DMSO or dimethyl formamide; soluble in 100% ethanol (50mg/ml); sparingly soluble in basic aqueous buffers (1.7mg/ml). To prevent oxidation, the solvent should be purged with an inert gas.
Vzorec: C₂₀H₃₂O₂
Storage Temperature: Freezer
MDL Number: MFCD00004417
CAS číslo: 506-32-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9fb4d76eb5d18774d8c797a11c39852cd40043b&vid=cc141ea3ea68f1ffa081b44c197170c1a3c74302&prid=web&clid=SAPDAM