2-Methoxyestradiol ≥95%
Dodavatel: ENZO LIFE SCIENCES
BMLS5400010
BMLS5400050
ENZOBMLS5400010EA
2480
CZK
ENZOBMLS5400010
ENZOBMLS5400050
2-Methoxyestradiol ≥95%
2-Methoxyestradiol
Estrogen receptor inhibitor
An endogenous estrogen metabolite with low affinity for the estrogen receptor but which disrupts microtubule function. Potent inhibitor of endothelial cell proliferation and migration. Inhibits bFGF and VEGF-induced corneal neovascularization in mice. Posttranscriptionally downregulates HIF-1alpha expression.
Vzorec:
C₁₉H₂₆O₃ |
MDL Number:
MFCD00010489 CAS číslo: 362-07-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...