Tisk…
2',5'-Dideoxyadenosine ≥98% (by TLC)
Dodavatel: ENZO LIFE SCIENCES
Synonyms: (2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-2-methyltetrahydrofuran-3-ol
BMLCN1100005 BMLCN1100025
ENZOBMLCN1100005EA 4710 CZK
ENZOBMLCN1100005 ENZOBMLCN1100025
2',5'-Dideoxyadenosine ≥98% (by TLC)
2',5'-Dideoxyadenosine
Adenylate cyclase inhibitor
A useful, cell-permeable inhibitor of adenylate cyclase (ID50=250µM). Inhibits growth and caues cell differentiation (1.5µM) in K562 cells.
Vzorec: C₁₀H₁₃N₅O₂
MW: 235,2425 g/mol
MDL Number: MFCD00210904
CAS číslo: 6698-26-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ab68638f143149a79ebd3bac821ea05e3418c80&vid=cc06f876d932bc8df02eac72521b6066164f9ad3&prid=web&clid=SAPDAM