Tisk…

Bepridil hydrochloride ≥98%

Dodavatel: ENZO LIFE SCIENCES

ALX550237M010 ALX550237M025
ENZOALX550237M010EA 1670 CZK
ENZOALX550237M010 ENZOALX550237M025
Bepridil hydrochloride ≥98%
Bepridil hydrochloride

Non-selective Ca²⁺ channel blocker. Stimulates the binding of 1,4-dihydropyridine-based drugs to Ca²⁺ channels. Inhibits Na⁺/Ca²⁺ exchange.

Storage Temperature: Ambient
CAS číslo: 74764-40-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR