Tisk…

Anti-Rabbit IgG Chicken Polyclonal Antibody (CF®583R)

Dodavatel: Biotium
20891-50UL 20891-500UL
20891-50ULEA 2350 CZK
20891-50UL 20891-500UL
Anti-Rabbit IgG Chicken Polyclonal Antibody (CF®583R)
Protilátky
This is a chicken anti-rabbit IgG (H L) secondary antibody labeled with superior CF® Dyes. CF® Dyes offer exceptional brightness and photostability.

Type: Secondary
Antigen: Rabbit IgG
Clonality: Polyclonal
Clone:
Conjugation: CF®583R
Epitope:
Host: Chicken
Isotype: IgG2a (H+L)
Reactivity: Rabbit
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR