Tisk…

Anti-IgG Goat Polyclonal Antibody (CF®583R)

Dodavatel: Biotium
20899-50UL 20899-500UL
20899-50ULEA 2350 CZK
20899-50UL 20899-500UL
Anti-IgG Goat Polyclonal Antibody (CF®583R)
Protilátky
Goat anti-guinea pig IgG (H L) secondary antibody labelled with our superior CF® Dyes.

  • Suitable for western, immunofluorescence, and immunohistology in FFPE tissues
  • CF® Dye and biotin conjugates: add 0,5 ml dH₂O
  • Ex/Em: 585/609 nm

Add the indicated volume of water directly to the vial containing the lyophilised antibody and mix gently to dissolve. Store reconstituted antibody at −20 °C and protect from light. Aliquot to avoid repeated freeze/thaw cycles. Alternatively, an equal volume of glycerol can be mixed with the reconstituted antibody so that it will remain liquid at −20 °C.

Liquid format: Store at −20 °C, protected from light. Product is stable for at least 6 months from date of receipt when stored as recommended. Liquid format antibodies contain 50% glycerol and will not freeze at −20 °C.

Lyophilised format: Store at −20 °C, protected from light. Product is stable for at least 6 months from date of receipt when stored as recommended.

Upozornení: Storage of the antibody for more than a day at final working dilution is not recommended.

Type: Secondary
Antigen: IgG (Guinea pig)
Clonality: Polyclonal
Clone:
Conjugation: CF®583R
Epitope:
Host: Goat
Isotype:
Reactivity: Guinea pig
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR