Tisk…

Perfluorotriamylamine

Dodavatel: Merck

Synonyms: Perfluorotripentylamine

F9880-100ML F9880-25ML F9880-SPEC
SIALF9880-100MLEA 35770 CZK
SIALF9880-100ML SIALF9880-25ML SIALF9880-SPEC
Perfluorotriamylamine
Perfluorotriamylamine

Fluorinert™ FC-70 has been used to reduce the adherence of large trypsin:N-amidinopiperidine crystals to the well. It has also been used as a fluorocarbon oil for microinjection of oocytes. For use as a high density chase solution in density gradient studies.

Vzorec: C₁₅F₃₃N
MW: 821.12 g/mol
Hustota: 1.93 g/ml (25 °C) (lit.)
MDL Number: MFCD00042367
CAS číslo: 338-84-1
EINECS: 206-421-8

Objednat

Výsledky specifikačního testu

Appearance (Turbidity) Clear
Appearance (Colour) Colourless
Appearance (Form) Liquid
Boiling Point 212.0 - 218.0 °C
Density 1.910 - 1.950 g/ml
Dielectric Strength ≥35 kV
Distillation Range ≥95 %
Pour Point ≤−15 °C
Residue (100 ml) ≤10.0 µg/ml
Viscosity at 25.0 +/- 0.1 Degrees Celsius 11.0 - 17.0
Vendor Information Confirmed
Recommended Retest Period 3 Years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR