Tisk…
Cell Culture Media, Powdered
Dodavatel: Corning
50-003-PBT 50-003-PCT 50-003-PDT 50-010-PBT 50-010-PCT 50-010-PDT 50-011-PBT 50-011-PCT 50-011-PDT 50-020-PBT 50-020-PCT 50-020-PDT 51-010-PBT 51-010-PCT 51-010-PDT 58-003-PBT 58-003-PCT 58-003-PDT 58-007-PBT 58-007-PCT 58-007-PDT 58-008-PBT 58-008-PCT 58-008-PDT 58-009-PBT 58-009-PCT 58-009-PDT 58-011-PBT 58-011-PCT 58-011-PDT 58-012-PBT 58-012-PCT 58-012-PDT 58-014-PBT 58-014-PCT 58-014-PDT 58-015-PBT 58-015-PCT 58-015-PDT 58-021-PBT 58-021-PCT 58-021-PDT 58-022-PBT 58-022-PCT 58-022-PDT 58-031-PBT 58-031-PCT 58-031-PDT 58-042-PBT 58-042-PCT 58-042-PDT 58-043-PBT 58-043-PCT 58-043-PDT 58-044-PBT 58-044-PCT 58-044-PDT 90-020-PBT 90-020-PCT 90-020-PDT 90-025-PBT 90-025-PCT 90-025-PDT 90-093-PBT 90-093-PCT 90-093-PDT 90-095-PBT 90-095-PCT 90-095-PDT 90-103-PBT 90-103-PCT 90-103-PDT 90-104-PBT 90-104-PCT 90-104-PDT 90-105-PBT 90-105-PCT 90-105-PDT 90-106-PBT 90-106-PCT 90-106-PDT 90-021-PBT 90-021-PCT 90-021-PDT 90-026-PBT 90-026-PCT 90-026-PDT 90-102-PBT 90-102-PCT 90-102-PDT 90-040-PBT 90-040-PCT 90-040-PDT 90-041-PBT 90-041-PCT 90-041-PDT 90-010-PBT 90-010-PCT 90-010-PDT 90-030-PBT 90-030-PCT 90-030-PDT 90-031-PBT 90-031-PCT 90-031-PDT 90-032-PBT 90-032-PCT 90-032-PDT 90-033-PBT 90-033-PCT 90-033-PDT 90-051-PBT 90-051-PCT 90-051-PDT 90-060-PBT 90-060-PCT 90-060-PDT 90-070-PBT 90-070-PCT 90-070-PDT 90-071-PBT 90-071-PCT 90-071-PDT 90-080-PBT 90-080-PCT 90-080-PDT
MDTC50-003-PBTEA 651 CZK
MDTC50-003-PBT MDTC50-003-PCT MDTC50-003-PDT MDTC50-010-PBT MDTC50-010-PCT MDTC50-010-PDT MDTC50-011-PBT MDTC50-011-PCT MDTC50-011-PDT MDTC50-020-PBT MDTC50-020-PCT MDTC50-020-PDT MDTC51-010-PBT MDTC51-010-PCT MDTC51-010-PDT MDTC58-003-PBT MDTC58-003-PCT MDTC58-003-PDT MDTC58-007-PBT MDTC58-007-PCT MDTC58-007-PDT MDTC58-008-PBT MDTC58-008-PCT MDTC58-008-PDT MDTC58-009-PBT MDTC58-009-PCT MDTC58-009-PDT MDTC58-011-PBT MDTC58-011-PCT MDTC58-011-PDT MDTC58-012-PBT MDTC58-012-PCT MDTC58-012-PDT MDTC58-014-PBT MDTC58-014-PCT MDTC58-014-PDT MDTC58-015-PBT MDTC58-015-PCT MDTC58-015-PDT MDTC58-021-PBT MDTC58-021-PCT MDTC58-021-PDT MDTC58-022-PBT MDTC58-022-PCT MDTC58-022-PDT MDTC58-031-PBT MDTC58-031-PCT MDTC58-031-PDT MDTC58-042-PBT MDTC58-042-PCT MDTC58-042-PDT MDTC58-043-PBT MDTC58-043-PCT MDTC58-043-PDT MDTC58-044-PBT MDTC58-044-PCT MDTC58-044-PDT MDTC90-020-PBT MDTC90-020-PCT MDTC90-020-PDT MDTC90-025-PBT MDTC90-025-PCT MDTC90-025-PDT MDTC90-093-PBT MDTC90-093-PCT MDTC90-093-PDT MDTC90-095-PBT MDTC90-095-PCT MDTC90-095-PDT MDTC90-103-PBT MDTC90-103-PCT MDTC90-103-PDT MDTC90-104-PBT MDTC90-104-PCT MDTC90-104-PDT MDTC90-105-PBT MDTC90-105-PCT MDTC90-105-PDT MDTC90-106-PBT MDTC90-106-PCT MDTC90-106-PDT MDTC90-021-PBT MDTC90-021-PCT MDTC90-021-PDT MDTC90-026-PBT MDTC90-026-PCT MDTC90-026-PDT MDTC90-102-PBT MDTC90-102-PCT MDTC90-102-PDT MDTC90-040-PBT MDTC90-040-PCT MDTC90-040-PDT MDTC90-041-PBT MDTC90-041-PCT MDTC90-041-PDT MDTC90-010-PBT MDTC90-010-PCT MDTC90-010-PDT MDTC90-030-PBT MDTC90-030-PCT MDTC90-030-PDT MDTC90-031-PBT MDTC90-031-PCT MDTC90-031-PDT MDTC90-032-PBT MDTC90-032-PCT MDTC90-032-PDT MDTC90-033-PBT MDTC90-033-PCT MDTC90-033-PDT MDTC90-051-PBT MDTC90-051-PCT MDTC90-051-PDT MDTC90-060-PBT MDTC90-060-PCT MDTC90-060-PDT MDTC90-070-PBT MDTC90-070-PCT MDTC90-070-PDT MDTC90-071-PBT MDTC90-071-PCT MDTC90-071-PDT MDTC90-080-PBT MDTC90-080-PCT MDTC90-080-PDT
Cell Culture Media, Powdered
Média kultivační
Corning provides an extensive lineup of powdered basal and specialty media to complement liquid media offering including popular DMEM, MEM, and RPMI formulations, as well as specialised serum-free, serum-reduced, and virus maintenance media.
All powdered media are produced in a facility according to cGMP and ISO 13485 requirements to ensure high quality and reproducibility. In-house performance studies exhibit excellent cell density and viability.